Learn More
Alfa Aesar™ 2-Fluoro-N-n-pentylbenzamide, 97%
Brand: Alfa Aesar™ H59622.03
Code : NEW
Additional Details : CAS Number : 330469-11-9 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2-Fluoro-N-n-pentylbenzamide | |
330469-11-9 | |
209.26 | |
MFCD01214077 |
Specifications
2-Fluoro-N-n-pentylbenzamide | |
1g | |
Viscous Liquid | |
209.26 |
Pale yellow | |
330469-11-9 | |
97% | |
MFCD01214077 |