missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(5-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59881.03

Code : NEW

Additional Details : CAS Number : 710310-20-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466318

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(5-fluoro-2-methylphenyl)benzamide
710310-20-6
247.24
MFCD04067407

Specifications

2-Fluoro-N-(5-fluoro-2-methylphenyl)benzamide
710310-20-6
97%
MFCD04067407
1g
Solid
247.24
Videos
SDS