missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-5-(trifluoromethyl)benzamide, 95%

Brand:  Alfa Aesar™ H65612.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479776

  • £70.28 / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-5-(trifluoromethyl)benzamide
337.32
MFCD27975792

Specifications

2-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-5-(trifluoromethyl)benzamide
Solid
337.32
1g
95%
MFCD27975792
Videos
SDS