Learn More
Alfa Aesar™ 2-Fluoro-N-[3-(trifluoromethyl)phenyl]benzamide, 97%
Brand: Alfa Aesar™ H59052.03
Code : NEW
Additional Details : CAS Number : 2053-95-4 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2-Fluoro-N-[3-(trifluoromethyl)phenyl]benzamide | |
2053-95-4 | |
283.23 | |
MFCD00045081 |
Specifications
2-Fluoro-N-[3-(trifluoromethyl)phenyl]benzamide | |
2053-95-4 | |
97% | |
MFCD00045081 |
1g | |
Solid | |
283.23 |