missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(3-iodophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59429.03

Code : NEW

Additional Details : CAS Number : 443638-58-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465748

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(3-iodophenyl)benzamide
443638-58-2
341.12
MFCD02904294

Specifications

2-Fluoro-N-(3-iodophenyl)benzamide
1g
Solid
341.12
Light Sensitive
443638-58-2
97%
MFCD02904294
Videos
SDS