missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(3-fluoro-4-bromophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59338.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405618

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(3-fluoro-4-bromophenyl)benzamide
312.11
MFCD21525891

Specifications

2-Fluoro-N-(3-fluoro-4-bromophenyl)benzamide
Solid
312.11
1g
97%
MFCD21525891
Videos
SDS