missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(3-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59716.03

Code : NEW

Additional Details : CAS Number : 1283340-25-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496108

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(3-fluoro-2-methylphenyl)benzamide
1283340-25-9
247.24
MFCD16428102

Specifications

2-Fluoro-N-(3-fluoro-2-methylphenyl)benzamide
1283340-25-9
97%
MFCD16428102
1g
Solid
247.24
Videos
SDS