missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(2-fluoro-4-chlorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59048.03

Code : NEW

Additional Details : CAS Number : 545440-44-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15435228

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(2-fluoro-4-chlorophenyl)benzamide
545440-44-6
267.66
MFCD03391782

Specifications

2-Fluoro-N-(2-fluoro-4-chlorophenyl)benzamide
545440-44-6
97%
MFCD03391782
1g
Solid
267.66
Videos
SDS