missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N-(2,4-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59373.03

Code : NEW

Additional Details : CAS Number : 331435-72-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15435668

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-N-(2,4-difluorophenyl)benzamide
331435-72-4
251.21
MFCD00120214

Specifications

2-Fluoro-N-(2,4-difluorophenyl)benzamide
331435-72-4
97%
MFCD00120214
1g
Solid
251.21
Videos
SDS