missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog Number 10786275
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
32890-94-1
C8H3F4O2
207.10
MFCD00040981
LNARMXLVVGHCRP-UHFFFAOYSA-M
2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626
688123
[O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F
This item is not returnable. View return policy
32890-94-1
C8H3F4O2
207.10
MFCD00040981
LNARMXLVVGHCRP-UHFFFAOYSA-M
2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626
688123
[O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point 85.0°C to 87.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H3(CF3)CO2H
Quantity 1 g
IUPAC Name 2-fluoro-6-(trifluoromethyl)benzoate
Formula Weight 208.11
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-6-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'