missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Boiling Point | 156°C |
Flash Point | 82°C |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Formula Weight | 192.11 |
Percent Purity | 98% |
Chemical Name or Material | 2-Fluoro-6-(trifluoromethyl)benzaldehyde |
Safety and Handling
Product Identifier
- 2-Fluoro-6-(trifluoromethyl)benzaldehyde
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'