missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-nitrobenzyl bromide, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 50°C to 54°C |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | O2NC6H3CH2Br(F) |
Quantity | 1 g |
Synonym | 2-fluoro-6-nitrobenzyl bromide, 2-bromomethyl-1-fluoro-3-nitrobenzene, benzene, 2-bromomethyl-1-fluoro-3-nitro, alpha-bromo-2-fluoro-6-nitrotoluene, acmc-20anuk, 2-fluoro-6-nitrobenzylbromide, 2-bromomethyl-3-fluoro-1-nitrobenzene, benzene,2-bromomethyl-1-fluoro-3-nitro |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 234.02 |
Percent Purity | 98% |
Chemical Name or Material | 2-Fluoro-6-nitrobenzyl bromide |
Safety and Handling
Product Identifier
- 2-Fluoro-6-nitrobenzyl bromide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'