missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-nitrobenzyl bromide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Melting Point | 50°C to 54°C |
Odor | Pungent |
Quantity | 1 g |
UN Number | UN3261 |
Beilstein | 2503670 |
Synonym | 2-fluoro-6-nitrobenzyl bromide, 2-bromomethyl-1-fluoro-3-nitrobenzene, benzene, 2-bromomethyl-1-fluoro-3-nitro, alpha-bromo-2-fluoro-6-nitrotoluene, acmc-20anuk, 2-fluoro-6-nitrobenzylbromide, 2-bromomethyl-3-fluoro-1-nitrobenzene, benzene,2-bromomethyl-1-fluoro-3-nitro |
Solubility Information | Insoluble in water. |
Formula Weight | 234.02 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Fluoro-6-nitrobenzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'