missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7052669
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
117752-04-2
C8H7FO
138.14
MFCD09258962
BGQBJWPZBBMKEI-UHFFFAOYSA-N
14170517
2-fluoro-6-methylbenzaldehyde
CC1=C(C=O)C(F)=CC=C1
This item is not returnable. View return policy
117752-04-2
C8H7FO
138.14
MFCD09258962
BGQBJWPZBBMKEI-UHFFFAOYSA-N
14170517
2-fluoro-6-methylbenzaldehyde
CC1=C(C=O)C(F)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.151 g/mL
Refractive Index 1.525
Quantity 1 g
Synonym 2-fluoro-6-methyl-benzaldehyde, benzaldehyde,2-fluoro-6-methyl, pubchem10067, 3-fluoro-2-formyltoluene, acmc-209xa9, benzaldehyde, 2-fluoro-6-methyl, 2-fluoro-6-methylbenzaldehyde
Formula Weight 138.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-methylbenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'