Alfa Aesar™ 2-Fluoro-6-methylbenzaldehyde, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H55228.06

5GR 2-Fluoro-6-methylbenzaldehyde, 97%

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 117752-04-2 Weight: 0.00500kg


Quantity 5g

Product Code. 15441408

Quantity Price
1 £ 256.52 / 5g
Estimated Shipment date
from Supplier 09-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Fluoro-6-methylbenzaldehyde
CAS 117752-04-2
Formula Weight 138.14
MDL Number MFCD09258962

Specification

Formula Weight 138.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Fluoro-6-methylbenzaldehyde
Refractive Index 1.5250
Physical Form Liquid
Assay 97%
MDL Number MFCD09258962
CAS 117752-04-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Density 1.151g/mL
Color Colorless to yellow
Quantity 5g
Safety and Handling

Warning

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.