missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™

Catalog Number p-4228670
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
137654-21-8
C8H7FO3
170.14
MFCD00671764
XLNQGZLCGVLNMF-UHFFFAOYSA-N
2-fluoro-6-methoxy-benzoic acid, rarechem al bo 0442, benzoic acid, 2-fluoro-6-methoxy, pubchem4967, acmc-209cem, 6-fluoro-2-anisoic acid, intermediates-zcf02063, methyl-6-fluorosalicylic acid, ksc494o0r, 2-fluoro-6-methoxybenzoicacid
2737360
2-fluoro-6-methoxybenzoic acid
COC1=C(C(=CC=C1)F)C(=O)O
This item is not returnable. View return policy
137654-21-8
C8H7FO3
170.14
MFCD00671764
XLNQGZLCGVLNMF-UHFFFAOYSA-N
2-fluoro-6-methoxy-benzoic acid, rarechem al bo 0442, benzoic acid, 2-fluoro-6-methoxy, pubchem4967, acmc-209cem, 6-fluoro-2-anisoic acid, intermediates-zcf02063, methyl-6-fluorosalicylic acid, ksc494o0r, 2-fluoro-6-methoxybenzoicacid
2737360
2-fluoro-6-methoxybenzoic acid
COC1=C(C(=CC=C1)F)C(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 85°C to 87°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 170.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-methoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-6-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'