missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzoyl chloride, 97%, ACROS Organics™

Brand:  Acros Organics 426130010

Code : 29

Additional Details : CAS Number : 111771-12-1 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10537444

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Fluoro-6-iodobenzoyl chloride
96.0
97%
FC6H3(I)COCl
MFCD03094410
111771-12-1
100.0
C7H3ClFIO
284.45

Specifications

2-Fluoro-6-iodobenzoyl chloride
1g
96.0
Glass bottle
C7H3ClFIO
284.45
1.956g/mL
111771-12-1
100.0
97%
FC6H3(I)COCl
MFCD03094410
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Fluoro-6-iodobenzoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS