missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)phenol, 98%, Thermo Scientific™

Catalog Number p-3759056
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
141483-15-0
C7H4F4O
180.10
MFCD00061286
MCOSBFKOUQAIJS-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl phenol, 4-fluoro-3-hydroxybenzotrifluoride, 2-fluoro-5-trifluoromethyl-phenol, phenol, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-m-cresol, pubchem4145, intermediates-zcf02079, acmc-1c6g6, ksc495m9p, 5-trifluoromethyl-2-fluorophenol
518888
2-fluoro-5-(trifluoromethyl)phenol
OC1=CC(=CC=C1F)C(F)(F)F
This item is not returnable. View return policy
141483-15-0
C7H4F4O
180.10
MFCD00061286
MCOSBFKOUQAIJS-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl phenol, 4-fluoro-3-hydroxybenzotrifluoride, 2-fluoro-5-trifluoromethyl-phenol, phenol, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-m-cresol, pubchem4145, intermediates-zcf02079, acmc-1c6g6, ksc495m9p, 5-trifluoromethyl-2-fluorophenol
518888
2-fluoro-5-(trifluoromethyl)phenol
OC1=CC(=CC=C1F)C(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Orange
Density 1.4360g/mL
Boiling Point 146.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(OH)CF3
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.436
Formula Weight 180.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluoro-5-(trifluoromethyl)phenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'