missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-nitropyridine, 98%, Thermo Scientific Chemicals
102.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Melting Point | 17°C to 21°C |
Color | Colorless to Yellow |
Density | 1.41 g/mL |
Boiling Point | 86°C to 87°C (7 mmHg)(literature) |
Refractive Index | 1.525 |
Quantity | 25 g |
Beilstein | 120351 |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 142.09 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'