missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-nitrobenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Formula Weight | 166.11 |
Percent Purity | 97% |
Chemical Name or Material | 2-Fluoro-5-nitrobenzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'