missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylaniline, 99%, Thermo Scientific™

Catalog Number 10510892
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 452-84-6
CAS Min % 98.5
CAS Max % 100.0
Density 1.1090g/mL
Boiling Point 80°C to 86°C (11.0mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8FN
Linear Formula FC6H3(CH3)NH2
Refractive Index 1.5320 to 1.534
MDL Number MFCD00007654
Quantity 5g
Specific Gravity 1.109
Synonym 3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
Solubility Information Solubility in water:
InChI Key QZUXMXZNVAJNSE-UHFFFAOYSA-N
SMILES CC1=CC=C(F)C(N)=C1
IUPAC Name 2-fluoro-5-methylaniline
Molecular Weight (g/mol) 125.15
PubChem CID 262970
Formula Weight 125.15
Percent Purity 99%
Chemical Name or Material 2-Fluoro-5-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-5-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'