missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylaniline, 99%, ACROS Organics™

Brand:  Acros Organics 276150250

Code : 29

Additional Details : CAS Number : 452-84-6 Weight : 0.08000kg Transport : UN number : 2810 Chem class : 1 Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10058541

  • £75.00 / 25g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Fluoro-5-methylaniline
98.5
≥98.5%
C7H8FN
125.15
2-Fluoro-5-methylbenzeneamine
452-84-6
100.0
99%
FC6H3(CH3)nH2
MFCD00007654

Specifications

2-Fluoro-5-methylaniline
Clear yellow to brown
452-84-6
100.0
Glass bottle
1.5320 to 1.5340 (20°C, 589nm)
≥98.5%
C7H8FN
125.15
2-Fluoro-5-methylbenzeneamine
1.109g/mL
25g
98.5
Authentic
Product darkens on storage
Liquid
99%
FC6H3(CH3)nH2
MFCD00007654
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Fluoro-5-methylaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS