missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine, 97%, Thermo Scientific™

Catalog Number 15498665
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1435806-24-8
C11H11F4NO2
265.208
MFCD19687203
ZAEBGVLZTPXBJV-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl-dl-phenylalanine, 2-amino-3-2-fluoro-5-methyl-4-trifluoromethyl phenyl propanoic acid
66523503
2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid
CC1=CC(=C(C=C1C(F)(F)F)F)CC(C(=O)O)N
This item is not returnable. View return policy
1435806-24-8
C11H11F4NO2
265.208
MFCD19687203
ZAEBGVLZTPXBJV-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl-dl-phenylalanine, 2-amino-3-2-fluoro-5-methyl-4-trifluoromethyl phenyl propanoic acid
66523503
2-amino-3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid
CC1=CC(=C(C=C1C(F)(F)F)F)CC(C(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 265.2
Percent Purity 97%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'