missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™

Catalog Number 15478875
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1373920-64-9
C10H7F4NO
233.17
MFCD22201045
PRSLVVFXWPHVSP-UHFFFAOYSA-N
2-fluoro-5-methoxy-3-trifluoromethyl phenylacetonitrile, 2-2-fluoro-5-methoxy-3-trifluoromethyl phenyl acetonitrile
86277646
2-[2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]acetonitrile
COC1=CC(=C(F)C(CC#N)=C1)C(F)(F)F
This item is not returnable. View return policy
1373920-64-9
C10H7F4NO
233.17
MFCD22201045
PRSLVVFXWPHVSP-UHFFFAOYSA-N
2-fluoro-5-methoxy-3-trifluoromethyl phenylacetonitrile, 2-2-fluoro-5-methoxy-3-trifluoromethyl phenyl acetonitrile
86277646
2-[2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]acetonitrile
COC1=CC(=C(F)C(CC#N)=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 233.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'