missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(hydroxymethyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7030511
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
1072952-25-0
C7H8BFO3
169.95
MFCD06656276
CMJGYVJNQJJJOZ-UHFFFAOYSA-N
2-fluoro-5-hydroxymethylphenylboronic acid, 2-fluoro-5-hydroxymethyl phenyl boronic acid, 2-fluoro-5-hydroxymethyl phenylboronic acid, 3-borono-4-fluorobenzyl alcohol, 2-fluoro-5-hydroxymethylphenylbornic acid, 2-fluoro-5-hydroxymethyl benzeneboronic acid, pubchem5040, acmc-2098uj, ksc494m6l
44717544
[2-fluoro-5-(hydroxymethyl)phenyl]boronic acid
OCC1=CC(B(O)O)=C(F)C=C1
This item is not returnable. View return policy
1072952-25-0
C7H8BFO3
169.95
MFCD06656276
CMJGYVJNQJJJOZ-UHFFFAOYSA-N
2-fluoro-5-hydroxymethylphenylboronic acid, 2-fluoro-5-hydroxymethyl phenyl boronic acid, 2-fluoro-5-hydroxymethyl phenylboronic acid, 3-borono-4-fluorobenzyl alcohol, 2-fluoro-5-hydroxymethylphenylbornic acid, 2-fluoro-5-hydroxymethyl benzeneboronic acid, pubchem5040, acmc-2098uj, ksc494m6l
44717544
[2-fluoro-5-(hydroxymethyl)phenyl]boronic acid
OCC1=CC(B(O)O)=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 64°C to 76°C
Quantity 1 g
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-(hydroxymethyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'