missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-formylbenzonitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-3771013
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
218301-22-5
C8H4FNO
149.12
MFCD01863558
MOFRJTLODZILCR-UHFFFAOYSA-N
3-cyano-4-fluorobenzaldehyde, 4-fluoro-3-cyanobenzaldehyde, 2-fluoro-5-formyl-benzonitrile, benzonitrile, 2-fluoro-5-formyl, 2-fluoro-5-formylbenzenecarbonitrile, 2-fluor-5-formylbenzonitrile, 10014 2-fluoro-5-formyl benzonitrile, pubchem2252, acmc-1cfa9, fluoro-5-formylbenzonitrile
2769582
2-fluoro-5-formylbenzonitrile
C1=CC(=C(C=C1C=O)C#N)F
This item is not returnable. View return policy
218301-22-5
C8H4FNO
149.12
MFCD01863558
MOFRJTLODZILCR-UHFFFAOYSA-N
3-cyano-4-fluorobenzaldehyde, 4-fluoro-3-cyanobenzaldehyde, 2-fluoro-5-formyl-benzonitrile, benzonitrile, 2-fluoro-5-formyl, 2-fluoro-5-formylbenzenecarbonitrile, 2-fluor-5-formylbenzonitrile, 10014 2-fluoro-5-formyl benzonitrile, pubchem2252, acmc-1cfa9, fluoro-5-formylbenzonitrile
2769582
2-fluoro-5-formylbenzonitrile
C1=CC(=C(C=C1C=O)C#N)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80°C to 84°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H3(CN)CHO
Quantity 1 g
Formula Weight 149.12
Percent Purity 99%
Chemical Name or Material 2-Fluoro-5-formylbenzonitrile
compliance-icons
Product Identifier
  • 2-Fluoro-5-formylbenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'