missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62689.06

Code : NEW

Additional Details : CAS Number : 1112208-82-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15477736

  • £406.10 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-5-formylbenzeneboronic acid pinacol ester
250.07
2-Fluoro-5-formylphenylboronic acid pinacol ester
1112208-82-8
MFCD11867877

Specifications

2-Fluoro-5-formylbenzeneboronic acid pinacol ester
5g
Solid
250.07
2-Fluoro-5-formylphenylboronic acid pinacol ester
35° to 40°C
1112208-82-8
96%
MFCD11867877
Videos
SDS