missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-5-(3-methylphenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H54903.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15401746

  • £271.00 / 1g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1449135-55-0
273.07
DULJUFCUHPACGN-UHFFFAOYSA-N
73996021
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC(=C2)C)F)(O)O
C14H13BFNO3
MFCD20265368
2-fluoro-5-3-methylphenylcarbamoyl benzeneboronic acid, 2-fluoro-5-3-methylphenyl carbamoyl phenylboronic acid, 2-fluoro-5-m-tolylcarbamoyl phenylboronic acid
[2-fluoro-5-[(3-methylphenyl)carbamoyl]phenyl]boronic acid

Specifications

2-Fluoro-5-(3-methylphenylcarbamoyl)benzeneboronic acid
1449135-55-0
MFCD20265368
DULJUFCUHPACGN-UHFFFAOYSA-N
[2-fluoro-5-[(3-methylphenyl)carbamoyl]phenyl]boronic acid
73996021
97%
1g
C14H13BFNO3
2-fluoro-5-3-methylphenylcarbamoyl benzeneboronic acid, 2-fluoro-5-3-methylphenyl carbamoyl phenylboronic acid, 2-fluoro-5-m-tolylcarbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC=CC(=C2)C)F)(O)O
273.07
273.07
Videos
SDS Documents
Certificates