missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15467756
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1073353-68-0
C13H15BF4O2
290.06
MFCD08458187
ZXEYGQNXUCXTBK-UHFFFAOYSA-N
2-fluoro-4-trifluoromethylphenylboronic acid, pinacol ester, 2-2-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl-phenylboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl benzeneboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl phenyl boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-2-fluoro-4-trifluoromethyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
17750248
2-[2-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
1073353-68-0
C13H15BF4O2
290.06
MFCD08458187
ZXEYGQNXUCXTBK-UHFFFAOYSA-N
2-fluoro-4-trifluoromethylphenylboronic acid, pinacol ester, 2-2-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl-phenylboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl benzeneboronic acid pinacol ester, 2-fluoro-4-trifluoromethyl phenyl boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-2-fluoro-4-trifluoromethyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
17750248
2-[2-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'