missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number 15427776
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1073353-89-5
C12H15BFNO4
267.063
MFCD09264075
QYXHQOSFZITWSU-UHFFFAOYSA-N
2-2-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-nitrophenylboronic acid, pinacol ester, 2-fluoro-4-nitrophenylboronic acid,pinacol ester
46738935
2-(2-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)[N+](=O)[O-])F
This item is not returnable. View return policy
1073353-89-5
C12H15BFNO4
267.063
MFCD09264075
QYXHQOSFZITWSU-UHFFFAOYSA-N
2-2-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-nitrophenylboronic acid, pinacol ester, 2-fluoro-4-nitrophenylboronic acid,pinacol ester
46738935
2-(2-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)[N+](=O)[O-])F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 267.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-nitrobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'