missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-4-methyl-3-nitropyridine, 98%

Brand:  Alfa Aesar™ H64837.MD

Code : NEW

Additional Details : CAS Number : 19346-43-1 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15492219

  • £9.80 / 250mg


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-4-methyl-3-nitropyridine
156.12
1639010
19346-43-1
MFCD04112526

Specifications

2-Fluoro-4-methyl-3-nitropyridine
250mg
Solid
156.12
1639010
33°C
19346-43-1
98%
MFCD04112526
Videos
SDS