missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-4-iodobenzonitrile, 98%

Brand:  Alfa Aesar™ H61973.03

Code : NEW

Additional Details : CAS Number : 137553-42-5 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15448568

  • £42.60 / 1g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Description

Description

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

2-Fluoro-4-iodobenzonitrile
247.01
UN3439
137553-42-5
MFCD03094172

Specifications

2-Fluoro-4-iodobenzonitrile
Red-brown
137553-42-5
98%
MFCD03094172
Light Sensitive
1g
Powder
247.01
UN3439
Videos
SDS