missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog Number 15468568
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
137553-42-5
C7H3FIN
247.011
MFCD03094172
WHQVXHBSTRFRCE-UHFFFAOYSA-N
3-fluoro-4-cyano-1-iodobenzene, benzonitrile, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenecarbonitrile, pubchem4778, acmc-209xcp, intermediates-zcf02061, 4-iodo-2-fluorobenzonitrile, 2-fluoro-4-iodo-benzonitrile, ksc493i5t, benzonitrile,2-fluoro-4-iodo
2774518
2-fluoro-4-iodobenzonitrile
C1=CC(=C(C=C1I)F)C#N
This item is not returnable. View return policy
137553-42-5
C7H3FIN
247.011
MFCD03094172
WHQVXHBSTRFRCE-UHFFFAOYSA-N
3-fluoro-4-cyano-1-iodobenzene, benzonitrile, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenecarbonitrile, pubchem4778, acmc-209xcp, intermediates-zcf02061, 4-iodo-2-fluorobenzonitrile, 2-fluoro-4-iodo-benzonitrile, ksc493i5t, benzonitrile,2-fluoro-4-iodo
2774518
2-fluoro-4-iodobenzonitrile
C1=CC(=C(C=C1I)F)C#N

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color Red to Brown
Quantity 250 mg
UN Number UN3439
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 247.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-4-iodobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'