missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-4-iodo-5-methylpyridine, 95%

Brand:  Alfa Aesar™ H64983.03

Code : NEW

Additional Details : CAS Number : 153034-94-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402429

  • £65.20 / 1g

Estimated Shipment: 27-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

153034-94-7
237.016
BVKQLNXPPQEELX-UHFFFAOYSA-N
10060052
CC1=CN=C(C=C1I)F
C6H5FIN
MFCD03095297
2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
2-fluoro-4-iodo-5-methylpyridine

Specifications

2-Fluoro-4-iodo-5-methylpyridine
White
1g
C6H5FIN
6643371
BVKQLNXPPQEELX-UHFFFAOYSA-N
2-fluoro-4-iodo-5-methylpyridine
10060052
Solid
Light sensitive
40°C to 45°C
153034-94-7
MFCD03095297
2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
CC1=CN=C(C=C1I)F
237.016
237.02
95%
Videos
SDS Documents
Certificates