missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7036931
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1449144-94-8
C13H9BF2N2O5
322.031
MFCD20040308
JSRCTYHYPFPKJW-UHFFFAOYSA-N
2-fluoro-4-4-fluoro-2-nitrophenylcarbamoyl benzeneboronic acid, 2-fluoro-4-4-fluoro-2-nitrophenyl carbamoyl phenylboronic acid
73996130
[2-fluoro-4-[(4-fluoro-2-nitrophenyl)carbamoyl]phenyl]boronic acid
B(C1=C(C=C(C=C1)C(=O)NC2=C(C=C(C=C2)F)[N+](=O)[O-])F)(O)O
This item is not returnable. View return policy
1449144-94-8
C13H9BF2N2O5
322.031
MFCD20040308
JSRCTYHYPFPKJW-UHFFFAOYSA-N
2-fluoro-4-4-fluoro-2-nitrophenylcarbamoyl benzeneboronic acid, 2-fluoro-4-4-fluoro-2-nitrophenyl carbamoyl phenylboronic acid
73996130
[2-fluoro-4-[(4-fluoro-2-nitrophenyl)carbamoyl]phenyl]boronic acid
B(C1=C(C=C(C=C1)C(=O)NC2=C(C=C(C=C2)F)[N+](=O)[O-])F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 322.03
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'