missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-nitro-6-methylpyridine, 98%

Brand:  Alfa Aesar™ H64103.06

Code : NEW

Additional Details : CAS Number : 19346-45-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15461149

  • £167.60 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Description

Description

Applications
6-Methyl-3-nitro-2-(2-phenylhydrazinyl)pyridine (MNPHP) was synthesized from 2-fluoro-3-nitro-6-methylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

2-Fluoro-3-nitro-6-methylpyridine
156.12
1638807
19346-45-3
MFCD03095067
2-Fluoro-3-nitro-6-picoline

Specifications

2-Fluoro-3-nitro-6-methylpyridine
30°C
19346-45-3
98%
MFCD03095067
2-Fluoro-3-nitro-6-picoline
Off-white
5g
Solid
156.12
1638807
Videos
SDS