missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 95%, Thermo Scientific™

Catalog Number p-7052845
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
MFCD28098710
This item is not returnable. View return policy
MFCD28098710

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 196.97
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-(methylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'