Learn More
Thermo Scientific Maybridge 2-Fluoro-3-iodopyridine, 5g, 97%
Brand: Thermo Scientific Maybridge MO00359DE
2-Fluoro-3-iodopyridine, 5g, 97%
Code : 31
Additional Details : CAS Number : 113975-22-7 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge MO00359DE
2-Fluoro-3-iodopyridine, 5g, 97%
Code : 31
Additional Details : CAS Number : 113975-22-7 Weight : 0.05500kg