missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-(3-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56699.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402846

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-3-(3-fluorobenzylcarbamoyl)benzeneboronic acid
291.06
MFCD22682883

Specifications

2-Fluoro-3-(3-fluorobenzylcarbamoyl)benzeneboronic acid
Solid
291.06
1g
97%
MFCD22682883
Videos
SDS