missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-[2-(trifluoromethyl)phenylcarbamoyl]benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56621.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15492806

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-3-[2-(trifluoromethyl)phenylcarbamoyl]benzeneboronic acid
327.04
MFCD22682877

Specifications

2-Fluoro-3-[2-(trifluoromethyl)phenylcarbamoyl]benzeneboronic acid
Solid
327.04
1g
97%
MFCD22682877
Videos
SDS