missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56423.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15422686

  • £11.98 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid
260.03

Specifications

2-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid
Solid
260.03
1g
97%
Videos
SDS