missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-(2-methoxyphenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56889.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432956

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-3-(2-methoxyphenylcarbamoyl)benzeneboronic acid
289.07
MFCD22682894

Specifications

2-Fluoro-3-(2-methoxyphenylcarbamoyl)benzeneboronic acid
1g
97%
MFCD22682894
Off-white
Solid
289.07
Videos
SDS