missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-(2-methoxybenzylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56279.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402596

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-3-(2-methoxybenzylcarbamoyl)benzeneboronic acid
303.1
MFCD22682859

Specifications

2-Fluoro-3-(2-methoxybenzylcarbamoyl)benzeneboronic acid
1g
97%
MFCD22682859
Off-white
Solid
303.1
Videos
SDS