missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-(2-fluorophenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56684.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15442836

  • £358.49 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluoro-3-(2-fluorophenylcarbamoyl)benzeneboronic acid
277.03
MFCD22682881

Specifications

2-Fluoro-3-(2-fluorophenylcarbamoyl)benzeneboronic acid
Solid
277.03
1g
97%
MFCD22682881
Videos
SDS