missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Ethylthiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62626.03

Code : NEW

Additional Details : CAS Number : 14347-73-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409138

  • £109.18 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Ethylthiazolidine-4-carboxylic acid
14347-73-0
161.22
MFCD00185986

Specifications

2-Ethylthiazolidine-4-carboxylic acid
14347-73-0
97%
MFCD00185986
1g
Solid
161.22
Videos
SDS