missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Ethylthiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62626.06

Code : NEW

Additional Details : CAS Number : 14347-73-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419138

  • £543.78 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Ethylthiazolidine-4-carboxylic acid
14347-73-0
161.22
MFCD00185986

Specifications

2-Ethylthiazolidine-4-carboxylic acid
14347-73-0
97%
MFCD00185986
5g
Solid
161.22
Videos
SDS