missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylpyrrole, 98%, Thermo Scientific Chemicals

Catalog Number p-3757904
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1551-06-0
C6H9N
95.15
MFCD00060477
XRPDDDRNQJNHLQ-UHFFFAOYSA-N
137075
2-ethyl-1H-pyrrole
CCC1=CC=CN1
This item is not returnable. View return policy
1551-06-0
C6H9N
95.15
MFCD00060477
XRPDDDRNQJNHLQ-UHFFFAOYSA-N
137075
2-ethyl-1H-pyrrole
CCC1=CC=CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 0.9290g/mL
Boiling Point 142.0°C (15.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4985 to 1.5005
Quantity 250 mg
Specific Gravity 0.929
Synonym 2-ethylpyrrole, 1h-pyrrole, 2-ethyl, pyrrole,2-ethyl, 1h-pyrrole,2-ethyl, acmc-209dbz, #, 2-ethylpyrrole, analytical standard, 2-ethylpyrrole, technical grade
Formula Weight 95.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethylpyrrole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'