missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylimidazole, 99%, ACROS Organics™

£24.10 - £252.79

Chemical Identifiers

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-ethyl-1H-imidazole
SMILES CCC1=NC=CN1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10185080 SDS View Documents Acros Organics
219281000
100g Glass bottle £24.10
100g View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Please to purchase this item. Need a web account? Register with us today!
10042860 SDS View Documents Acros Organics
219285000
500g Glass bottle £77.41
500g View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Please to purchase this item. Need a web account? Register with us today!
10434525 SDS View Documents Acros Organics
219280025
2.5kg Plastic Drum £252.79
2.50kg View alternative products
Save Up to 
»
Estimated Shipment
24-08-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1072-62-4
96.13
PQAMFDRRWURCFQ-UHFFFAOYSA-N
66130
CCC1=NC=CN1
C5H8N2
MFCD00005192
2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
2-ethyl-1H-imidazole

Specifications

2-Ethylimidazole
Glass bottle
83.0°C to 87.0°C
98%
98.5
1% max. Imidazole (GC)
MFCD00005192
2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PQAMFDRRWURCFQ-UHFFFAOYSA-N
2-ethyl-1H-imidazole
66130
99%
159°C
268.0°C
100g
1072-62-4
100.0
C5H8N2
23, 78
Solubility in water: 617g/L (20°C). Other solubilities: soluble in many organic solvents
CCC1=NC=CN1
96.13
96.13
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Ethylimidazole

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.