missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3598163
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
688-84-6
C12H22O2
198.31
MFCD00009494
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
12713
2-ethylhexyl 2-methylprop-2-enoate
CCCCC(CC)COC(=O)C(=C)C
This item is not returnable. View return policy
688-84-6
C12H22O2
198.31
MFCD00009494
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
12713
2-ethylhexyl 2-methylprop-2-enoate
CCCCC(CC)COC(=O)C(=C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Density 0.8840g/mL
Boiling Point 120°C (18.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH(C2H5)(CH2)3CH3
Refractive Index 1.4370 to 1.4390
Quantity 1 L
Specific Gravity 0.884
Solubility Information Solubility in water: 0.0031g/L
Stabilizer 40 to 60ppm MEHQ
Formula Weight 198.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl methacrylate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethylhexyl methacrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'