Learn More
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
Description
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.
Solubility
Miscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Melting Point | -59°C |
Density | 0.91 |
pH | 3 |
Boiling Point | 226°C to 229°C |
Flash Point | 114°C (237°F) |
Refractive Index | 1.425 |
Quantity | 100 mL |
Beilstein | 1750468 |
Solubility Information | Miscible with water. |
Formula Weight | 144.21 |
Show More |
RUO – Research Use Only